Learn More
MSC Misty Carpet Cleaner, Liquid, Pleasant Scent, 1 gal Bottle
Catalog No. 502838581
Encompass
This concentrated, low-foaming shampoo is designed for cleaning carpets, rugs, and upholstery. It can be used with cold- or hot-water extraction machines or steam cleaning units, effectively cleaning water-soluble soils and oil-based matter such as grease and oils. The formula includes an odor counteractant to keep the area smelling fresh.
  • Concentrated, low-foaming formula.
  • Compatible with cold or hot-water extraction machines and steam cleaning units.
  • Effectively cleans water-soluble soils, oil-based matter, grease, and oils.
  • Contains an odor counteractant for a pleasant and fresh scent.
  • Suitable for use on natural and synthetic textiles, carpets, and upholstery.
  • Ships as a case of four 1 gal bottles.
Catalog No. 50-283-8581 Supplier MSC Supplier No. 50539980
This concentrated, low-foaming shampoo is designed for cleaning carpets, rugs, and upholstery. It can be used with cold- or hot-water extraction machines or steam cleaning units, effectively cleaning water-soluble soils and oil-based matter such as grease and oils. The formula includes an odor counteractant to keep the area smelling fresh.
  • Concentrated, low-foaming formula.
  • Compatible with cold or hot-water extraction machines and steam cleaning units.
  • Effectively cleans water-soluble soils, oil-based matter, grease, and oils.
  • Contains an odor counteractant for a pleasant and fresh scent.
  • Suitable for use on natural and synthetic textiles, carpets, and upholstery.
  • Ships as a case of four 1 gal bottles.