Learn More
PDC Misys/Sunquest Direct Thermal Paper Label, 1200 Labels/Roll, 8 Rolls/Case
Catalog No. 21235808
Change view
Click to view available options
Quantity:
8 Rolls/Cs.
Catalog No. Quantity
21-235-808 8 Rolls/Cs.
1 options
Catalog No. 21-235-808 Supplier PDC Supplier No. LDWHP97
Add to Cart
Add to Cart
Provide permanent adhesion and sharp, clear printing, ideal for lab and clinical specimen labeling.
  • Direct thermal paper labels with permanent adhesive for clinical and lab use
  • White face stock for clear, easy-to-read printing
  • Designed for specimen and patient identification
  • Core fits compatible dispensers for easy handling
  • Compatible with Misys and Sunquest EHR systems
  • Bulk supply for high-volume labeling needs
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 8 Rolls/Cs.
Dimensions (L x W) 4 1/8 x 1 3/16 in.
Length (English) 4.125 in.
Width (English) 1.1875 in.
Width (Metric) 30.1625 mm