Learn More
PDC Misys/Sunquest/Epic Direct Thermal Paper Label, White,1000 Labels/Roll, 6 Rolls/Box
Catalog No. 21235825
Change view
Click to view available options
Quantity:
1000 Labels/Roll, 6 Rolls/Box
Catalog No. Quantity
21-235-825 1000 Labels/Roll, 6 Rolls/Box
1 options
Catalog No. 21-235-825 Supplier PDC Supplier No. TD1SQ4
  • $147.00
    $109.65 / Case of 6
    Save $37.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Direct thermal labels compatible with Misys, Sunquest, and Epic, glove-friendly with permanent adhesive for reliable specimen labeling.
  • Designed for efficient workflow in clinical and laboratory environments
  • Medical-grade quality to minimize labeling errors
  • Supports clear text and accurate barcodes for specimen tracking
  • Durable materials suitable for frequent handling
  • Easy-to-use format for high-volume labeling operations
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1000 Labels/Roll, 6 Rolls/Box
Dimensions (L x W) 4 1/8 x 1 3/16 in.
Length (English) 4.125 in.
Width (English) 1.1875 in.
Width (Metric) 30.1625 mm