Learn More
Apexbio Technology LLC Mitochondrial Membrane Potential and Apoptosis Detection Kit 50T
SDP
Catalog No. 503107184
Encompass_Preferred
Mito-Tracker Red CMXRos (MitoMark Red I) is a red fluorescent oxidation-sensitive probe that specifically labels mitochondria It can permeate live cell membranes and selectively accumulate in mitochondria When mitochondrial membrane potential decreases the intensity of red fluorescence also diminishes thus enabling evaluation of mitochondrial damage based on fluorescence intensity Mitochondrial Membrane Potential and Apoptosis Detection Kit is designed to simultaneously detect apoptosis and changes in mitochondrial membrane potential by employing Annexin V-FITC (a green-fluorescence apoptosis probe) and Mito-Tracker Red CMXRos Additionally the kit includes Hoechst 33342 which stains cell nuclei Following staining live cells appear positive for red fluorescence and negative for green fluorescence while apoptotic cells appear positive for green fluorescence and exhibit significantly reduced or negative red fluorescence
Catalog No. 50-310-7184 Supplier Apexbio Technology LLC Supplier No. K226450
Mito-Tracker Red CMXRos (MitoMark Red I) is a red fluorescent oxidation-sensitive probe that specifically labels mitochondria It can permeate live cell membranes and selectively accumulate in mitochondria When mitochondrial membrane potential decreases the intensity of red fluorescence also diminishes thus enabling evaluation of mitochondrial damage based on fluorescence intensity Mitochondrial Membrane Potential and Apoptosis Detection Kit is designed to simultaneously detect apoptosis and changes in mitochondrial membrane potential by employing Annexin V-FITC (a green-fluorescence apoptosis probe) and Mito-Tracker Red CMXRos Additionally the kit includes Hoechst 33342 which stains cell nuclei Following staining live cells appear positive for red fluorescence and negative for green fluorescence while apoptotic cells appear positive for green fluorescence and exhibit significantly reduced or negative red fluorescence