Learn More
Medchemexpress LLC Mitapivat | 1260075-17-9 | 100 MG
SDP
Catalog No. 5000339033
Encompass_Preferred
Mitapivat is an orally active pyruvate kinase allosteric activator used in the study of PK deficiency.
  • Increases enzymatic activity
  • Increases protein stability
  • Increases ATP levels across a wide range of PKLR genotypes
  • Shows potential to restore the activity of PK (pyruvate kinase)-deficient glycolytic pathways
Catalog No. 50-003-39033 Supplier Medchemexpress LLC Supplier No. HY12689100MG
Mitapivat is an orally active pyruvate kinase allosteric activator used in the study of PK deficiency.
  • Increases enzymatic activity
  • Increases protein stability
  • Increases ATP levels across a wide range of PKLR genotypes
  • Shows potential to restore the activity of PK (pyruvate kinase)-deficient glycolytic pathways