Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitchell Plastics Beta Storage Containers

Catalog No. 22312443
Change view
Click to view available options
Height (Metric):
12cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
22-312443 12cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-312443 Supplier Mitchell Plastics Supplier No. RP300

missing translation for 'validMainframeMsg'

Safely store and transport reagents that emit beta isotopes

  • Safely store reagents that emit beta isotopes including 3H, 14C, 32P and 35S
  • 0.953cm (0.375 in.) thick acrylic offers protection from beta radiation exposure during transport, storage or general use
  • Withstands temperatures as low as -70°C (-94°F)
  • Hinged lids provide easy interior access
  • Dimensions: 20.3L x 28W x 12.1cmH (8 x 11 x 4.75H in.)

Specifications

Product Type Beta storage container
Includes Hinged lid
For Use With (Application) For safe storage of reagents that emit beta isotopes such as 3H, 14C, 32P, and 35S
Length (English) 8 in.
Length (Metric) 20.3cm
Width (Metric) 28cm
Width (English) 11 in.
Dimensions (L x W x H) 8 x 11 x 4.75 in. (20.3 x 28 x 12cm)
Thickness (English) 0.37 in.
Height (Metric) 12cm
Height (English) 4.75 in.
Material Acrylic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.