Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitchell Plastics Biohazard Benchtop Waste Containers

Catalog No. 05723435
Change view
Click to view available options
Height (English):
8.5 in.
15.5 in.
Length (English):
4 in.
7.25 in.
Width (English):
4.5 in.
8.5 in.
For Use With (Equipment):
20.32 x 30.48 cm Bag
35.56 x 48.26 cm Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Length (English) Width (English) For Use With (Equipment)
05-723-435 15.5 in. 7.25 in. 8.5 in. 35.56 x 48.26 cm Bag
05-723-424 8.5 in. 4 in. 4.5 in. 20.32 x 30.48 cm Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 05-723-435 Supplier Mitchell Plastics Supplier No. WC300

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Clear containers facilitate easy and safe segregation of hazardous materials for later disposal

  • Made of heavy-duty acrylic to help protect personnel from exposure to potential hazards
  • Clear design enables warning insignias and biohazard bag colors to be seen
  • Each unit has a hinged lid for easy placement of waste
  • Lids are also removable for easy positioning and removal of bag or box 
  • Reusable

 

Bench top models

  • WC-300: holds bag 35.56 × 48.26cm (14 × 19 in.)
  • WC-400: holds bag 20.32 × 30.48cm (8 × 12 in.)
  • WB-600: holds glass disposal box 20.32 × 20.32 × 25.4cm (8 × 8 × 10 in.)

Specifications

Color Clear
Dimensions (L x W x H) 7.25 x 8.5 x 15.5 in.
Height (Metric) 39.3 cm
Length (Metric) 18.42 cm
Model Benchtop
Size Large
Width (Metric) 21.6 cm
For Use With (Application) Waste disposal
For Use With (Equipment) 35.56 x 48.26 cm Bag
Width (English) 8.5 in.
Length (English) 7.25 in.
Thickness 4.75 mm
Height (English) 15.5 in.
Material Plastic
Product Type Benchtop Biohazard Waste Container
Show More Show Less

Dispose contents according to all local, state and federal disposal regulations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.