Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitchell Plastics Refrigerator Storage Box with Lock

Catalog No. 05723471
Change view
Click to view available options
Clear/Green; 3.75 x 9.5 x 6.625 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
05-723-471 Clear/Green; 3.75 x 9.5 x 6.625 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-723-471 Supplier Mitchell Plastics Supplier No. SBCLCG396

missing translation for 'validMainframeMsg'

Store drugs, reagents, liquids and other contents from unauthorized users with the electronic, touch pad lock or combination locks.

  • Combination lock provides for use by authorized users without having to locate and share a key
  • Space savings size to full size to accommodate whatever the need
  • Combination locks can be easily programmable
  • Keyed locks available
  • Removable drawer for easy re-filling
  • 3/16 in. thick acrylic provides protection from exposure to beta radiation during transport, storage or general use
  • Solid White Sintra with black drawer available to protect contents from view
  • Top of case has four slots for securing to cabinet shelves or racks in refrigerator
  • Can withstand temperatures as low as -70 ° C
  • Electronic, touch pad digital lock and combination locks
  • Select and change combination easily
  • Full size or space savings size to accommodate whatever the need
  • White, heavy-duty, ABS plastic or black acrylic to conceal contents
  • Security hinged door with security screws
  • 3/16 in. thick acrylic provides protection from exposure to beta radiation during transport, storage or general use
  • Double sided tape also provided to secure to glass shelves
  • Custom sizes are available

Specifications

Type Storage Box
Color Clear, Neon Green
Dimensions (L x W x H) 3.75 x 9.5 x 6.625 in.
For Use With (Application) Store Reagents, Liquids and Other Contents from Unauthorized Users with Lockable Storage Boxes
For Use With (Equipment) Cabinets, Shelves or Racks in Refrigeration Unit
Height (English) 3.75 in.
Includes Pull-Out Drawer
Length (English) 9.5 in.
Material Acrylic
Shape Rectangular
Width (English) 6.625 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.