Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitchell Plastics Re-usable, Heavy Duty Waste Containers, Floor Models

Catalog No. 05723432
Change view
Click to view available options
Height (Metric):
52.70 cm
63.5 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
05-723-432 52.70 cm
05-723-431 63.5 cm
05-723-434 63.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 05-723-432 Supplier Mitchell Plastics Supplier No. WC200
This item is not returnable. View return policy

Specifications

Depth (English) 19.75 in.
Closure Type Hinged Lid
Color Clear
Height (Metric) 52.70 cm
Width (Metric) 27.94 cm
For Use With (Application) Ideal for keeping sharp objects and liquid waste contained in the event of waste box leakage, disposing of biohazardous, as well as some radioactive materials
Width (English) 11 in.
Height (English) 20.75 in.
Material Acrylic
Product Type Floor Model Waste Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.