Learn More
Aviva Systems Biology Corp MITD1 Antibody - middle region (ARP52647_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52647_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MITD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MITD1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-746