Learn More
Aviva Systems Biology Corp MITF Antibody - N-terminal region (ARP37977_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37977_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MITF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MITF. Alternate names: MI, WS2, WS2A, bHLHe32ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56470