Learn More
Aviva Systems Biology Corp Mitf Antibody - middle region (ARP37860_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37860_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mitf. It was validated on Western Blot. Mitf,microphthalmia-associated transcription factor,Wh; bw; mi; vit; Bhlhe32; Vitiligo; MGC124309; MGC124310; Mitf,microphthalmia-associated transcription factor,microphthalmia-associated transcription factor,black eyed white,OTTMUSP00000027091,OTTMUSP00000027092The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: MGC124309, MGC124310, bw, mi, vit, vitiligo, wh, Wh, Bhlhe32, VitiligoApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-284-693