Learn More
TARGETMOL CHEMICALS INC MITHRAMYCIN A 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T55305MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mithramycin A (Plicamycin) a DNA-binding antitumor antibiotic is a selective specificity protein 1 (Sp1) inhibitor which inhibits the growth of various cancers by decreasing Sp1 protein. Specificity protein 1 (Sp1) is a zinc-finger transcription factor that regulates multiple cellular functions and promotes tumor progression. purity: 99%
Catalog No. 50-257-9749