Learn More
Abcam Mithramycin A (Plicamycin), DNA and RNA polymerase inhibitor, 5MG
Catalog No. 502706198
Encompass_Preferred
MW 1085.1 Da, Purity >98%. DNA and RNA polymerase inhibitor, DNA binding transcriptional inhibitor (Sp1 and Sp3). Shows antineoplastic activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6198 Supplier Abcam Supplier No. AB1427235MG
MW 1085.1 Da, Purity >98%. DNA and RNA polymerase inhibitor, DNA binding transcriptional inhibitor (Sp1 and Sp3). Shows antineoplastic activity.
The product is subject to the following: Abcam Restricted Use Statement