Learn More
Medchemexpress LLC HY-17398 100mg Medchemexpress, Mitiglinide (Calcium) CAS:145525-41-3 Purity:>98%
SDP
Catalog No. 502081943
Encompass_Preferred
Medchemexpress, HY-17398 100mg Mitiglinide (Calcium) CAS:145525-41-3 Mitiglinide Calcium (KAD-1229; S21403) is an ATP-sensitive K+ (KATP) channel antagonist. Mitiglinide Calcium is highly specific to the Kir6.2/SUR1 complex (the pancreatic beta-cell KATP channel). Mitiglinide Calcium can be used for the research of type 2 diabetes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1943 Supplier Medchemexpress LLC Supplier No. HY17398100MG
Medchemexpress, HY-17398 100mg Mitiglinide (Calcium) CAS:145525-41-3 Mitiglinide Calcium (KAD-1229; S21403) is an ATP-sensitive K+ (KATP) channel antagonist. Mitiglinide Calcium is highly specific to the Kir6.2/SUR1 complex (the pancreatic beta-cell KATP channel). Mitiglinide Calcium can be used for the research of type 2 diabetes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.