Learn More
Apexbio Technology LLC Mitiglinide Calcium, 500mg. Cas: 145525-41-3 MFCD: MFCD09955408. Potassium channel inhibitor
SDP
Supplier:  Apexbio Technology LLC B2193500
Encompass_Preferred
Mitiglinide is a drug for the treatment of type 2 diabetes. It is a highly selective KATP channel antagonist. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5329