Learn More
Selleck Chemical LLC Mitiglinide Calcium S2073-50mg
SDP
Catalog No. 503111269
Encompass_Preferred
Mitiglinide Calcium (KAD-1229) is a blood glucose-lowering drugs stimulating insulin secretion by closing the ATP-sensitive K channels in pancreatic beta-cells
Catalog No. 50-311-1269 Supplier Selleck Chemical LLC Supplier No. S207350MG
Mitiglinide Calcium (KAD-1229) is a blood glucose-lowering drugs stimulating insulin secretion by closing the ATP-sensitive K channels in pancreatic beta-cells