Learn More
Cayman Chemical MItIglInIdecalcIum salt 50mg
SDP
Catalog No. 502988757
Encompass_Preferred
A SUR1/Kir6.2 inhibitor (IC50 = 4 nM); selective for SUR1/Kir6.2 over SUR2A/Kir6.2 and SUR2B/Kir6.2 channels (IC50s = 3 and 5 µM, respectively); induces insulin release in HIT-T15 insulinoma cells and isolated mouse pancreatic islets at 100 µM; increases postprandial plasma insulin levels and inhibits postprandial increases in plasma glucose levels in an STZ-induced rat model of diabetes
Catalog No. 50-298-8757 Supplier Cayman Chemical Supplier No. 3069350 MG
A SUR1/Kir6.2 inhibitor (IC50 = 4 nM); selective for SUR1/Kir6.2 over SUR2A/Kir6.2 and SUR2B/Kir6.2 channels (IC50s = 3 and 5 µM, respectively); induces insulin release in HIT-T15 insulinoma cells and isolated mouse pancreatic islets at 100 µM; increases postprandial plasma insulin levels and inhibits postprandial increases in plasma glucose levels in an STZ-induced rat model of diabetes