Learn More
TARGETMOL CHEMICALS INC MITIPERSTAT 5MG
SDP
Catalog No. 502579558
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mitiperstat is a potent myeloperoxidase (MPO) inhibitor that is effective in preventing cardiovascular diseases such as heart failure and coronary artery disease. purity: 98%
Catalog No. 50-257-9558 Supplier TARGETMOL CHEMICALS INC Supplier No. T610285MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mitiperstat is a potent myeloperoxidase (MPO) inhibitor that is effective in preventing cardiovascular diseases such as heart failure and coronary artery disease. purity: 98%