Learn More
Apexbio Technology LLC MitMAB 1119-97-7 10mg
SDP
Supplier:  Apexbio Technology LLC B762010
Encompass_Preferred
MitMAB (CAS 1119-97-7) is a small-molecule inhibitor targeting dynamin a GTPase enzyme essential for membrane fission during endocytosis It is designed to interfere with dynamin-dependent endocytic processes by disrupting the molecular organization of membrane lipids and interacting with lipid bilayers MitMAB exerts its biological activity primarily through altering membrane lipid organization thereby inhibiting dynamin-mediated membrane trafficking Based on these pharmacological properties MitMAB holds research potential in the investigation of receptor internalization pathways analysis of viral entry mechanisms and studies of cellular membrane trafficking events
Catalog No. 50-224-0522