Learn More
Apexbio Technology LLC MITO DNA ISOLATION KIT-1mg
SDP
Catalog No. 5000897366
Encompass_Preferred
Mitochondrial DNA Isolation Kit is an essential tool in biomedical research enabling the extraction of high-purity mitochondrial DNA from various biological samples Originating from advancements in molecular biology this kit utilizes a targeted mechanism of action to isolate mitochondrial DNA by disrupting cell membranes and separating mitochondria through centrifugation It demonstrates efficient in vitro activity across a range of concentrations allowing researchers to obtain DNA in nanomolar to low micromolar dimensions Typical applications include analyzing mitochondrial diseases conducting cell model experiments and facilitating drug screening in animal studies Experimental concentrations typically depend on the specific research design This versatile kit supports the exploration of mitochondrial dysfunctions energy production processes and genetic studies enhancing our understanding of cellular metabolism and its implications in disease states
Catalog No. 50-008-97366 Supplier Apexbio Technology LLC Supplier No. K21171
Mitochondrial DNA Isolation Kit is an essential tool in biomedical research enabling the extraction of high-purity mitochondrial DNA from various biological samples Originating from advancements in molecular biology this kit utilizes a targeted mechanism of action to isolate mitochondrial DNA by disrupting cell membranes and separating mitochondria through centrifugation It demonstrates efficient in vitro activity across a range of concentrations allowing researchers to obtain DNA in nanomolar to low micromolar dimensions Typical applications include analyzing mitochondrial diseases conducting cell model experiments and facilitating drug screening in animal studies Experimental concentrations typically depend on the specific research design This versatile kit supports the exploration of mitochondrial dysfunctions energy production processes and genetic studies enhancing our understanding of cellular metabolism and its implications in disease states