Learn More
Dojindo Molecular Technologies Inc Mito-FerroGreen (1 set 50UG x2) is a novel fluorescent probe for the detection of ferrous ions (Fe2+) in mitochondria, used in ferroptosis research.
SDP
Catalog No. NC1631648
Encompass
Iron is the most abundant transition metal element within an organism and exhibits various physiological activities. Free iron in living cells is particularly important because of its high reactivity, suggesting a relationship to cellular damage or death. Free iron exists in its stable redox states, ferrous ion (Fe2+) and ferric ion (Fe3+). In living cells, research involving the behavior of Fe2+ is necessary to understand the intracellular reductive environment, metal transporters and water solubility of Fe2+. Mito-FerroGreen is a novel fluorescent probe for the detection of ferrous ion (Fe2+) in mitochondria where Fe-S clusters and heme proteins are synthesized, and enables live cell fluorescent imaging of intracellular Fe2+.Mito-FerroGreen is widely used in ferroptosis research, one of the various research fields Dojindo has a strong portfolio in. (https://www.dojindo.com/ferroptosis-detection-selection-guide/)
Catalog No. NC1631648 Supplier Dojindo Molecular Technologies Inc Supplier No. M48910
Iron is the most abundant transition metal element within an organism and exhibits various physiological activities. Free iron in living cells is particularly important because of its high reactivity, suggesting a relationship to cellular damage or death. Free iron exists in its stable redox states, ferrous ion (Fe2+) and ferric ion (Fe3+). In living cells, research involving the behavior of Fe2+ is necessary to understand the intracellular reductive environment, metal transporters and water solubility of Fe2+. Mito-FerroGreen is a novel fluorescent probe for the detection of ferrous ion (Fe2+) in mitochondria where Fe-S clusters and heme proteins are synthesized, and enables live cell fluorescent imaging of intracellular Fe2+.Mito-FerroGreen is widely used in ferroptosis research, one of the various research fields Dojindo has a strong portfolio in. (https://www.dojindo.com/ferroptosis-detection-selection-guide/)