Learn More
Selleck Chemical LLC Mitochondrial Fusion Promoter M1 S3375-5mg
SDP
Catalog No. 503112990
Encompass_Preferred
Mitochondrial fusion promoter M1 is a mitochondrial dynamic modulator that increases the expression of mitofusin 2 a mitochondrial outer membrane fusion protein reduces brain mitochondrial dysfunction Mitochondrial fusion promoter M1 alleviates brain damage in rats with cardiac ischemia/reperfusion injury
Catalog No. 50-311-2990 Supplier Selleck Chemical LLC Supplier No. S33755MG
Mitochondrial fusion promoter M1 is a mitochondrial dynamic modulator that increases the expression of mitofusin 2 a mitochondrial outer membrane fusion protein reduces brain mitochondrial dysfunction Mitochondrial fusion promoter M1 alleviates brain damage in rats with cardiac ischemia/reperfusion injury