Learn More
Enzo Life Sciences MITO-ID Membrane potential cytotoxicity kit (1x1Kit)
SDP
Catalog No. NC0100325
Encompass_Preferred
A Real-time Mitochondrial membrane Potential Assay with Superior Sensitivity. Applications: HTS, Microplate. Contents: MITO-ID MP Detection Reagent, 200 μL. CCCP Control, 100 μL . 10X Assay Buffer 1: 2.5 mL. 50X Assay Buffer 2: 0.5 mL
Catalog No. NC0100325 Supplier Enzo Life Sciences Supplier No. ENZ51019KP002
A Real-time Mitochondrial membrane Potential Assay with Superior Sensitivity. Applications: HTS, Microplate. Contents: MITO-ID MP Detection Reagent, 200 μL. CCCP Control, 100 μL . 10X Assay Buffer 1: 2.5 mL. 50X Assay Buffer 2: 0.5 mL