Learn More
Enzo Life Sciences MITO-ID Red detection kit (GFP-CERTIFIED) (500tests)
SDP
Catalog No. 502012582
Encompass_Preferred
MITO-ID Red Detection Kit (GFP-CERTIFIED) contains a proprietary membrane-permeable mitochondria-selective dye suitable for use with live-, detergent-permeabilized- and even aldehyde-fixed-cells. Unlike conventional dyes, such as DiOC6(3), JC-1, rhodamine 123 and tetramethylrhodamine ethyl ester, MITO-ID Red dye highlights mitochondria regardless of their energetic state. The dye is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes, as well as, High Content Screening (HCS) platforms. The kit is useful for assessing mitochondrial morphology changes, estimating mitochondrial mass and co-localizing GFP-tagged proteins to the mitochondrial compartment. Wavelength maxima: MITO-ID Red λex 558 nm, λem 690 nm; Hoechst 33342 λex 350 nm, λem 461 nm. Applications: Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-2582 Supplier Enzo Life Sciences Supplier No. ENZ51007500
MITO-ID Red Detection Kit (GFP-CERTIFIED) contains a proprietary membrane-permeable mitochondria-selective dye suitable for use with live-, detergent-permeabilized- and even aldehyde-fixed-cells. Unlike conventional dyes, such as DiOC6(3), JC-1, rhodamine 123 and tetramethylrhodamine ethyl ester, MITO-ID Red dye highlights mitochondria regardless of their energetic state. The dye is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes, as well as, High Content Screening (HCS) platforms. The kit is useful for assessing mitochondrial morphology changes, estimating mitochondrial mass and co-localizing GFP-tagged proteins to the mitochondrial compartment. Wavelength maxima: MITO-ID Red λex 558 nm, λem 690 nm; Hoechst 33342 λex 350 nm, λem 461 nm. Applications: Fluorescence microscopy, Fluorescent detection .