Learn More
Medchemexpress LLC Mito-LND (Mito-lonidamine) | 2361564-49-8 | 97.0% | 801.62 g/mol | C43H45BrCl2N3OP | 5 MG
SDP
Catalog No. 5000222913
Encompass_Preferred
Mito-LND (Mito-lonidamine) is a mitochondria-targeted inhibitor of oxidative phosphorylation used in research to probe mitochondrial bioenergetics and cancer metabolism. It reduces mitochondrial ATP production, increases reactive oxygen species, and can induce autophagic cell death in tumor models.
  • Mitochondria-targeted oxidative phosphorylation inhibitor.
  • Orally active research compound suitable for in vivo and in vitro studies.
  • Promotes reactive oxygen species formation and autophagic cell death.
  • Useful for studying mitochondrial bioenergetics and cancer metabolism.
  • Supplied as a solid powder with reported purity of 97.0%.
  • Chemical formula C43H45BrCl2N3OP; molar mass 801.62 g/mol.
Catalog No. 50-002-22913 Supplier Medchemexpress LLC Supplier No. HY1348325MG
Mito-LND (Mito-lonidamine) is a mitochondria-targeted inhibitor of oxidative phosphorylation used in research to probe mitochondrial bioenergetics and cancer metabolism. It reduces mitochondrial ATP production, increases reactive oxygen species, and can induce autophagic cell death in tumor models.
  • Mitochondria-targeted oxidative phosphorylation inhibitor.
  • Orally active research compound suitable for in vivo and in vitro studies.
  • Promotes reactive oxygen species formation and autophagic cell death.
  • Useful for studying mitochondrial bioenergetics and cancer metabolism.
  • Supplied as a solid powder with reported purity of 97.0%.
  • Chemical formula C43H45BrCl2N3OP; molar mass 801.62 g/mol.