Learn More
Apexbio Technology LLC MITO-TEMPO-100MG
SDP
Catalog No. 5000568901
Encompass_Preferred
Mito-TEMPO (CAS No 1334850-99-5) is a mitochondria-targeted antioxidant used in biomedical research Originating as a derivative of TEMPO it specifically targets mitochondrial reactive oxygen species (ROS) playing a crucial role in oxidative stress studies Its mechanism involves scavenging superoxide radicals thereby protecting against mitochondrial dysfunction Mito-TEMPO exhibits activity in the nanomolar to low micromolar range It is commonly applied in cell models and animal studies to investigate neurodegenerative diseases cardiovascular conditions and aging-related pathologies Drug screening often explores its potential therapeutic effects Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68901 Supplier Apexbio Technology LLC Supplier No. BA6176100
Mito-TEMPO (CAS No 1334850-99-5) is a mitochondria-targeted antioxidant used in biomedical research Originating as a derivative of TEMPO it specifically targets mitochondrial reactive oxygen species (ROS) playing a crucial role in oxidative stress studies Its mechanism involves scavenging superoxide radicals thereby protecting against mitochondrial dysfunction Mito-TEMPO exhibits activity in the nanomolar to low micromolar range It is commonly applied in cell models and animal studies to investigate neurodegenerative diseases cardiovascular conditions and aging-related pathologies Drug screening often explores its potential therapeutic effects Experimental concentrations typically depend on the specific research design