Learn More
Medchemexpress LLC Mito-TEMPO | 1334850-99-5 | 99.19% | 510.03 | 25 MG
SDP
Catalog No. 5000439573
Encompass_Preferred
Mito-TEMPO is a mitochondria-targeted superoxide dismutase mimetic with superoxide and alkyl radical scavenging properties.
  • Mito-TEMPO greatly attenuates the increase in ALT activities and reduces the areas of necrosis at both time points, indicating that the protection is sustained until at least 24 h post-APAP.
  • Mito-TEMPO could induce secondary apoptosis in the late phase of APAP hepatotoxicity.
  • Mito-TEMPO induces secondary apoptosis after APAP overdose by inhibition of RIP3.
Catalog No. 50-004-39573 Supplier Medchemexpress LLC Supplier No. 25MGHY112879
Mito-TEMPO is a mitochondria-targeted superoxide dismutase mimetic with superoxide and alkyl radical scavenging properties.
  • Mito-TEMPO greatly attenuates the increase in ALT activities and reduces the areas of necrosis at both time points, indicating that the protection is sustained until at least 24 h post-APAP.
  • Mito-TEMPO could induce secondary apoptosis in the late phase of APAP hepatotoxicity.
  • Mito-TEMPO induces secondary apoptosis after APAP overdose by inhibition of RIP3.