Learn More
TARGETMOL CHEMICALS INC MITO-TEMPO 25MG
SDP
Catalog No. 502581048
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mito-TEMPO is a mitochondria-targeted superoxide dismutase mimetic. Mito-TEMPO with superoxide and alkyl radical scavenging properties. purity: 98%
Catalog No. 50-258-1048 Supplier TARGETMOL CHEMICALS INC Supplier No. T1942825MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mito-TEMPO is a mitochondria-targeted superoxide dismutase mimetic. Mito-TEMPO with superoxide and alkyl radical scavenging properties. purity: 98%