Learn More
Enzo Life Sciences Mito-TEMPO, (5mg)
SDP
Catalog No. NC1037796
Encompass_Preferred
Mitochondria-targeted antioxidant. Mitochondria-targeted antioxidant with superoxide and alkyl radical scavenging properties, which may be used for both in vivo and in vitro experiments. Alternative Name: 2-(2,2,6,6-Tetramethylpiperidin-1-oxyl-4-ylamino)-2-oxoethyltriphenylphosphonium . chloride . monohydrate. Formula: C29H35N2O2ClP . H2O. MW: 510.1 . 18.0. Purity: ≥95% (TLC). Appearance: Colorless film to pale orange solid. Solubility: Soluble in water, DMSO, 100% ethanol or methanol. Use/Stability: Solutions in water, saline, ethanol or DMSO are stable at +4°C for 1-2 weeks. DMSO stock solution can be stored also at -20°C. Long Term Storage: -20°C
Catalog No. NC1037796 Supplier Enzo Life Sciences Supplier No. ALX430150M005
Mitochondria-targeted antioxidant. Mitochondria-targeted antioxidant with superoxide and alkyl radical scavenging properties, which may be used for both in vivo and in vitro experiments. Alternative Name: 2-(2,2,6,6-Tetramethylpiperidin-1-oxyl-4-ylamino)-2-oxoethyltriphenylphosphonium . chloride . monohydrate. Formula: C29H35N2O2ClP . H2O. MW: 510.1 . 18.0. Purity: ≥95% (TLC). Appearance: Colorless film to pale orange solid. Solubility: Soluble in water, DMSO, 100% ethanol or methanol. Use/Stability: Solutions in water, saline, ethanol or DMSO are stable at +4°C for 1-2 weeks. DMSO stock solution can be stored also at -20°C. Long Term Storage: -20°C