Learn More
Apexbio Technology LLC Mito-Tracker Red CMXRos 167095-09-2 50ug
SDP
Catalog No. 5000089408
Encompass_Preferred
Mito-Tracker Red CMXRos (CAS 167095-09-2) is a red-fluorescent probe targeting mitochondria It is designed to label and visualize mitochondria in live cells by selective accumulation thereby enabling the assessment of mitochondrial morphology and membrane potential Mito-Tracker Red CMXRos exerts its biological activity primarily through membrane potential-dependent uptake and covalent retention within mitochondria Based on these properties Mito-Tracker Red CMXRos holds research potential in mitochondrial morphology studies mitochondrial membrane potential assessments live-cell imaging apoptosis investigations and immunofluorescent microscopy
Catalog No. 50-000-89408 Supplier Apexbio Technology LLC Supplier No. B88100.05
Mito-Tracker Red CMXRos (CAS 167095-09-2) is a red-fluorescent probe targeting mitochondria It is designed to label and visualize mitochondria in live cells by selective accumulation thereby enabling the assessment of mitochondrial morphology and membrane potential Mito-Tracker Red CMXRos exerts its biological activity primarily through membrane potential-dependent uptake and covalent retention within mitochondria Based on these properties Mito-Tracker Red CMXRos holds research potential in mitochondrial morphology studies mitochondrial membrane potential assessments live-cell imaging apoptosis investigations and immunofluorescent microscopy