Learn More
Medchemexpress LLC MitoB bromide | 1247025-84-8 | MFCD28976194 | 99.8% | 477.14 g·mol⁻¹ | C25H23BBrO2P | 5 MG
SDP
Catalog No. 5000264550
Encompass_Preferred
MitoB bromide is a mitochondria-targeted ratiometric probe used as an exomarker to detect mitochondrial hydrogen peroxide (H2O2) in living animals. It accumulates in mitochondria and is converted by H2O2 to a reaction product that allows quantification of mitochondrial oxidative stress by analytical methods such as mass spectrometry.
  • Targeted detection of mitochondrial hydrogen peroxide in vivo.
  • Enables ratiometric quantification via conversion to a measurable product.
  • Compatible with mass spectrometry and analytical assays for in vivo studies.
  • Provided in research-scale quantities with high purity for reproducible results.
Catalog No. 50-002-64550 Supplier Medchemexpress LLC Supplier No. HY1264745MG
MitoB bromide is a mitochondria-targeted ratiometric probe used as an exomarker to detect mitochondrial hydrogen peroxide (H2O2) in living animals. It accumulates in mitochondria and is converted by H2O2 to a reaction product that allows quantification of mitochondrial oxidative stress by analytical methods such as mass spectrometry.
  • Targeted detection of mitochondrial hydrogen peroxide in vivo.
  • Enables ratiometric quantification via conversion to a measurable product.
  • Compatible with mass spectrometry and analytical assays for in vivo studies.
  • Provided in research-scale quantities with high purity for reproducible results.