Learn More
Medchemexpress LLC HY-115467 500ug Medchemexpress, MitoBloCK-10 CAS:394694-98-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY115467500UG
Encompass_Preferred
Medchemexpress, HY-115467 500ug MitoBloCK-10 CAS:394694-98-5 MitoBloCK-10 (MB-10) is the first small molecule modulator to attenuate protein-associated motor (PAM) complex activity. MitoBloCK-10 (MB-10) inhibits Tim44 (C-terminal domain) binding to the precursor and to Hsp70. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0881