Learn More
Cayman Chemical MitoBloCK-10, Purity: ≥98%, CAS: 394694-98-5, 50 mg
SDP
Catalog No. 502978463
Encompass_Preferred
A mitochondrial protein import inhibitor; inhibits the import of cytochrome c1 into the mitochondrial intermembrane space in yeast cells at 100 µM; inhibits migration and invasion of, and induces apoptosis and cell cycle arrest at the G1 phase in, primary human bladder cancer cells at 25 µM; induces mitochondrial depolarization, decreases ATP levels, and increases ROS in primary human bladder cancer cells; decreases tumor volume in a PDX mouse model of bladder cancer at 20 mg/kg every other day; intravitreal administration reduces SOD activity and increases TBARS levels and acellular capillary numbers in mouse retina at 0.25 nM/eye; induces dorsal curvature and cardiac apoptosis in zebrafish embryos at 10 µM
Catalog No. 50-297-8463 Supplier Cayman Chemical Supplier No. 4205450 MG
A mitochondrial protein import inhibitor; inhibits the import of cytochrome c1 into the mitochondrial intermembrane space in yeast cells at 100 µM; inhibits migration and invasion of, and induces apoptosis and cell cycle arrest at the G1 phase in, primary human bladder cancer cells at 25 µM; induces mitochondrial depolarization, decreases ATP levels, and increases ROS in primary human bladder cancer cells; decreases tumor volume in a PDX mouse model of bladder cancer at 20 mg/kg every other day; intravitreal administration reduces SOD activity and increases TBARS levels and acellular capillary numbers in mouse retina at 0.25 nM/eye; induces dorsal curvature and cardiac apoptosis in zebrafish embryos at 10 µM