Learn More
Dojindo Molecular Technologies Inc MitoBright ROS Deep Red, 3 x 100 nmol, Deep Red, 540 nm (λex), 670 nm (λem), High selectivity to superoxide, Ready to use, Fluorescence Microscopy, Flow Cytometry, Live Cell Imaging
SDP
Catalog No. NC2856087
Encompass
MitoBright ROS Deep Red, 3 x 100 nmol, Deep Red, 540 nm (λex), 670 nm (λem), High selectivity to superoxide, Ready to use, Fluorescence Microscopy, Flow Cytometry, Live Cell Imaging
Catalog No. NC2856087 Supplier Dojindo Molecular Technologies Inc Supplier No. MT1612
MitoBright ROS Deep Red, 3 x 100 nmol, Deep Red, 540 nm (λex), 670 nm (λem), High selectivity to superoxide, Ready to use, Fluorescence Microscopy, Flow Cytometry, Live Cell Imaging