Learn More
Cayman Chemical MItoCheck CItrate Synthase A
SDP
Catalog No. 502892319
Encompass_Preferred
The condensation of the dicarboxylate oxaloacetate and acetyl CoA to the tricarboxylate citrate is catalyzed by citrate synthase. It is within this reaction that carbon molecules (as acetyl CoA) obtained from pyruvate oxidation are fed into the tricarboxylic acid (TCA or citric acid) cycle. As a mitochondrial enzyme, citrate synthase is commonly used as a normalization factor for mitochondrial protein, but can also be used as a biomarker for mitochondrial content in a tissue homogenate. Cayman's MitoCheck Citrate Synthase Activity Assay Kit allows for the simple and convenient determination of citrate synthase activity from isolated mitochondria, tissue, or cell homogenates. This assay measures the production of SH-CoA by monitoring the absorbance of Citrate Synthase Developing Reagent at 412 nm in a convenient 96-well format.
Catalog No. 50-289-2319 Supplier Cayman Chemical Supplier No. 70104096 WELL
The condensation of the dicarboxylate oxaloacetate and acetyl CoA to the tricarboxylate citrate is catalyzed by citrate synthase. It is within this reaction that carbon molecules (as acetyl CoA) obtained from pyruvate oxidation are fed into the tricarboxylic acid (TCA or citric acid) cycle. As a mitochondrial enzyme, citrate synthase is commonly used as a normalization factor for mitochondrial protein, but can also be used as a biomarker for mitochondrial content in a tissue homogenate. Cayman's MitoCheck Citrate Synthase Activity Assay Kit allows for the simple and convenient determination of citrate synthase activity from isolated mitochondria, tissue, or cell homogenates. This assay measures the production of SH-CoA by monitoring the absorbance of Citrate Synthase Developing Reagent at 412 nm in a convenient 96-well format.