Learn More
Cayman Chemical MitoCheck Complex IV Activity Assay Kit, 96 wells
SDP
Supplier:  Cayman Chemical 70099096WELLS
Encompass_Preferred
Complex IV (cytochrome c oxidase) is the terminal electron acceptor in the mitochondrial electron transport chain. Complex IV functions by oxidizing cytochrome c and completing a four-electron reduction of oxygen (O2) to form water. During this process, two H+ are translocated from the mitochondrial matrix to the intermembrane space, contributing the mitochondrial membrane potential required for ATP synthesis. Common inhibitors of complex IV include potassium cyanide, azide, O2 limitation, nitric oxide, and carbon monoxide. Caymans MitoCheck Complex IV Activity Assay is designed to measure the direct oxidation of cytochrome c by complex IV, in an isolated bovine heart mitochondrial system that is supplied within the kit.
Catalog No. NC1402359