Learn More
Cayman Chemical MItoCheck Complex V ActIvIty
SDP
Catalog No. 502892244
Encompass_Preferred
An allosteric inhibitor of soluble adenylyl cyclase (IC50 50 11 UM) inhibits the activity of mitochondrial complex IV in wild-type but not sAC knockout MEFs at 50 UM reduces ischemia and reperfusion-induced swelling of and ATP content decreases in rat liver mitochondria at 1.2 mg/kg
Catalog No. 50-289-2244 Supplier Cayman Chemical Supplier No. 70100096 WELL
An allosteric inhibitor of soluble adenylyl cyclase (IC50 50 11 UM) inhibits the activity of mitochondrial complex IV in wild-type but not sAC knockout MEFs at 50 UM reduces ischemia and reperfusion-induced swelling of and ATP content decreases in rat liver mitochondria at 1.2 mg/kg