Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM2740P1
Encompass_Preferred
Product Type: Antibody. Application: IF, IHC. Conjugate/Tag: Unconjugated. Clone: MTC02. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HeLa or HepG2 cells. Hepatic carcinoma. Cellular Staining: Mitochondria in cytoplasm. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Semi-purified mitochondrial preparation. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C.
Catalog No. 50-188-5456