Learn More
Abcam Mitochondria/Cytosol Fractionation Kit, 100 Tests
Catalog No. 502204702
Encompass_Preferred
Mitochondria/cytosol fractionation kit provides a unique formulation of reagents for the effective isolation of highly enriched mitochondrial fractions from the cytosolic fraction of mammalian cells, including both apoptotic and non-apoptotic cells. - Cited in over 110 publications - Isolates mitochondrial and cytosolic fractions for studying apoptotic pathways and signal transduction - Simple and easy procedure for fractionation - Suitable for detecting translocation of factors between fractions via Western blotting (WB), ELISA, or other assays
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4702 Supplier Abcam Supplier No. AB65320100TEST
Mitochondria/cytosol fractionation kit provides a unique formulation of reagents for the effective isolation of highly enriched mitochondrial fractions from the cytosolic fraction of mammalian cells, including both apoptotic and non-apoptotic cells. - Cited in over 110 publications - Isolates mitochondrial and cytosolic fractions for studying apoptotic pathways and signal transduction - Simple and easy procedure for fractionation - Suitable for detecting translocation of factors between fractions via Western blotting (WB), ELISA, or other assays
The product is subject to the following: Abcam Restricted Use Statement