Learn More
Abcam Mitochondria Isolation Kit for Tissue, 1KIT
Catalog No. NC1912208
Encompass_Preferred
Mitochondria isolation kit for tissue is designed for the efficient isolation of mitochondria from hard or soft tissues, providing high-quality mitochondria for downstream analysis. The kit contains enough reagents for up to 10 isolations. - Cited in over 65 publications - Isolates mitochondria from both hard and soft tissues, such as brain, liver, and heart - Suitable for western blotting, immunocapture, or activity assays - Simple Protocol – no need for ultracentrifugation, making the process more efficient and user-friendly
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1912208 Supplier Abcam Supplier No. AB1101681KIT
Mitochondria isolation kit for tissue is designed for the efficient isolation of mitochondria from hard or soft tissues, providing high-quality mitochondria for downstream analysis. The kit contains enough reagents for up to 10 isolations. - Cited in over 65 publications - Isolates mitochondria from both hard and soft tissues, such as brain, liver, and heart - Suitable for western blotting, immunocapture, or activity assays - Simple Protocol – no need for ultracentrifugation, making the process more efficient and user-friendly
The product is subject to the following: Abcam Restricted Use Statement