Learn More
Abcam Mitochondria Isolation Kit for Cultured Cells, 1KIT
Catalog No. NC1481320
Encompass_Preferred
Mitochondria isolation kit for cultured cells is designed for efficient isolation of mitochondria from cultured mammalian cells. The kit provides enough reagents for up to 20 isolations and is suitable for analyzing mitochondria by Western blotting, immunocapture, or activity assays. - Cited in over 75 publications - Isolates mitochondria from cultured mammalian cells with ease - Useful in Western blotting, immunocapture, or mitochondrial activity assays - Sufficient reagents for up to 20 benchtop mitochondria isolations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1481320 Supplier Abcam Supplier No. AB1101701KIT
Mitochondria isolation kit for cultured cells is designed for efficient isolation of mitochondria from cultured mammalian cells. The kit provides enough reagents for up to 20 isolations and is suitable for analyzing mitochondria by Western blotting, immunocapture, or activity assays. - Cited in over 75 publications - Isolates mitochondria from cultured mammalian cells with ease - Useful in Western blotting, immunocapture, or mitochondrial activity assays - Sufficient reagents for up to 20 benchtop mitochondria isolations
The product is subject to the following: Abcam Restricted Use Statement