Learn More
Cayman Chemical MItochondrIal Complex II 10mL
SDP
Catalog No. 502892313
Encompass_Preferred
A G protein-biased agonist of GPR84 inhibits forskolin-induced cAMP production in CHO cells expressing human GPR84 (EC50 0.00598 nM) does not induce B-arrestin recruitment at concentrations up to 80 UM
Catalog No. 50-289-2313 Supplier Cayman Chemical Supplier No. 70094110 ML
A G protein-biased agonist of GPR84 inhibits forskolin-induced cAMP production in CHO cells expressing human GPR84 (EC50 0.00598 nM) does not induce B-arrestin recruitment at concentrations up to 80 UM