Learn More
Sigma Aldrich Fine Chemicals Biosciences Mitochondrial Complex III
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MAK3601KT
Encompass
Mitochondrial Complex III or Ubiquinol-Cytochrome c reductase is the third complex of the Electron Transport Chain located in the mitochondrial membrane. It is present in the mitochondria of all aerobic eukaryotes as well as in the inner membranes of most bacteria. It transfers electrons from CoQH (reduced coenzyme Q or ubiquinol) to cytochrome c resulting in the reduction of cytochrome c. This reduced cytochrome c is a substrate for complex IV of the electron transport chain.
Catalog No. 50-290-7220