Learn More
Cayman Chemical Mitochondrial Complex V ATP
SDP
Catalog No. 502894213
Encompass_Preferred
A Cdk4 inhibitor (Ki 0.6 nM for Cdk4/cyclin D1) selective for Cdk4 over Cdk6 (Ki 13.9 nM for Cdk6/cyclin D3) increases senescence in HR HER2- breast cancer cells in vitro but does not induce cell death in combination with fulvestrant
Catalog No. 50-289-4213 Supplier Cayman Chemical Supplier No. 7010044 MG
A Cdk4 inhibitor (Ki 0.6 nM for Cdk4/cyclin D1) selective for Cdk4 over Cdk6 (Ki 13.9 nM for Cdk6/cyclin D3) increases senescence in HR HER2- breast cancer cells in vitro but does not induce cell death in combination with fulvestrant