Learn More
Detroit R & D MITOCHONDRIAL DNA COPY
SDP
Catalog No. NC1125165
Encompass_Preferred
HUMAN MITOCHONDRIAL DNA COPY NUMBER. A quantitative PCR-based kit for measuring mitochondrial DNA copy number. This kit includes mouse or human or rat primers for an approximately 200 kb mitochondrial DNA sequence.
Catalog No. NC1125165 Supplier Detroit R & D Supplier No. MCN1
HUMAN MITOCHONDRIAL DNA COPY NUMBER. A quantitative PCR-based kit for measuring mitochondrial DNA copy number. This kit includes mouse or human or rat primers for an approximately 200 kb mitochondrial DNA sequence.