Learn More
Abcam Mitochondrial DNA Isolation Kit, 1KIT
Catalog No. 502215246
Encompass_Preferred
Mitochondrial DNA isolation kit provides efficient tools for isolating mitochondrial DNA (mtDNA) from a variety of cells and tissues, offering high yield and purity, free from genomic DNA contamination. - Cited in over 65 publications - Isolates mtDNA with no genomic DNA contamination - Suitable for enzyme manipulations, Southern blotting, PCR analysis, and amplifications - Simple and fast procedure for mtDNA isolation from cells or tissue - One kit is sufficient for 50 samples using adherent or suspension cells, and for 25 samples using tissue.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5246 Supplier Abcam Supplier No. AB653211KIT
Mitochondrial DNA isolation kit provides efficient tools for isolating mitochondrial DNA (mtDNA) from a variety of cells and tissues, offering high yield and purity, free from genomic DNA contamination. - Cited in over 65 publications - Isolates mtDNA with no genomic DNA contamination - Suitable for enzyme manipulations, Southern blotting, PCR analysis, and amplifications - Simple and fast procedure for mtDNA isolation from cells or tissue - One kit is sufficient for 50 samples using adherent or suspension cells, and for 25 samples using tissue.
The product is subject to the following: Abcam Restricted Use Statement