Learn More
Medchemexpress LLC Mitochondrial dye (MKT-077) | 147366-41-4 | MFCD31812307 | 98.5% | 432.00 g/mol | C21H22ClN3OS2 | 10 MG
SDP
Catalog No. 5000289965
Encompass_Preferred
MKT-077 is a cationic rhodacyanine mitochondrial dye and Hsp70 inhibitor used as a research reagent to probe mitochondrial function, induce apoptotic pathways, and study heat shock protein biology in vitro. It is provided as a high-purity powder for laboratory experiments.
  • Cationic rhodacyanine dye with Hsp70 inhibitory activity.
  • Used as a mitochondrial probe in apoptosis and cell viability studies.
  • High purity (98.49%) suitable for in vitro research.
  • Supplied as a 10 mg package for small-scale experiments.
  • Chemical formula C21H22ClN3OS2; molecular weight 432.00 g/mol.
  • Identifiers: CAS 147366-41-4, MFCD31812307.
This item is not returnable. View return policy
Catalog No. 50-002-89965 Supplier Medchemexpress LLC Supplier No. HY1509610MG
MKT-077 is a cationic rhodacyanine mitochondrial dye and Hsp70 inhibitor used as a research reagent to probe mitochondrial function, induce apoptotic pathways, and study heat shock protein biology in vitro. It is provided as a high-purity powder for laboratory experiments.
  • Cationic rhodacyanine dye with Hsp70 inhibitory activity.
  • Used as a mitochondrial probe in apoptosis and cell viability studies.
  • High purity (98.49%) suitable for in vitro research.
  • Supplied as a 10 mg package for small-scale experiments.
  • Chemical formula C21H22ClN3OS2; molecular weight 432.00 g/mol.
  • Identifiers: CAS 147366-41-4, MFCD31812307.