Learn More
Abcam Total OXPHOS Rodent WB Antibody Cocktail
Supplier:  Abcam AB110413
Encompass_Preferred
otal OXPHOS Rodent WB Antibody Cocktail ab110413 is an optimized cocktail of high quality antibodies for analyzing relative levels of OXPHOS complexes in rat or mouse mitochondria by western blot.- Measure the levels of OXPHOS complexes I through V, at the same time, in the same lane of a western blot- Use 5x less sample and take 5x less time than running separate western blots- Cited in over 450 publications
Catalog No. NC3779936