Learn More
Cayman Chemical MItochondrIal FIsIon Factr
SDP
Catalog No. 502938290
Encompass_Preferred
A naturally occurring saturated fatty acid inhibits lysis of human erythrocytes induced by S aureus delta toxin in a concentration-dependent manner decreased in the sebum of patients with papulopustular rosacea increased in malignant but not adjacent nonmalignant prostate tissue in patients with prostate cancer has been used as an internal standard for the quantification of fatty acids in chicken egg yolk
Catalog No. 50-293-8290 Supplier Cayman Chemical Supplier No. 29284100 UL
A naturally occurring saturated fatty acid inhibits lysis of human erythrocytes induced by S aureus delta toxin in a concentration-dependent manner decreased in the sebum of patients with papulopustular rosacea increased in malignant but not adjacent nonmalignant prostate tissue in patients with prostate cancer has been used as an internal standard for the quantification of fatty acids in chicken egg yolk